Stampa…

Magnesio acetato tetraidrato ≥98% ACS, Fluka™

Fornitore: Honeywell Chemicals

Synonyms: Acetic acid magnesium salt tetrahydrate, Magnesium di(acetate) tetrahydrate

228648-2.5KG 228648-500G 228648-10KG 228648-17.5KG
228648-2.5KGEA 540.54 EUR
228648-2.5KG 228648-500G HONC228648-10KG HONC228648-17.5KG
Magnesio acetato tetraidrato ≥98% ACS, Fluka™
Magnesio acetato tetraidrato
Formula: (H₃CCOO)₂Mg·4H₂O
Punto di fusione: 68 °C
Densità: 1,45 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149214
CAS Number: 16674-78-5
EINECS: 205-554-9
Merck Index: 13,05676

Visualizza i codici

Risultati del test di specificazione

Assay 98.0 - 102.0 %
Insoluble in acetic acid Max. 0.005 %
Barium (Ba) Max. 0.001 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.005 %
Manganese (Mn) Max. 0.001 %
Sodium (Na) Max. 0.005 %
Strontium (Sr) Max. 0.005 %
Heavy metals (ICP) ACS Max. 5 ppm
Chloride (Cl) Max. 0.001 %
Sulfate (SO₄) Max. 0.005 %
Form Solid


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR