Stampa…
Pump tubing, PharMed® BPT
Fornitore: Saint Gobain Life Sciences
070539-03 070539-08 070539-09 070539-14 070539-15 070539-16 070539-18 070539-19 070540-07 070540-08 070540-13 070540-15 070540-18 070539-07F 070539-09F 070539-10F 070539-12F 070539-19F 070539-13F 070539-14F 070539-15F 070539-16F 070539-18F 070540-05 070540-10 070540-11 070540-12 070540-19 070540-17 070539-03F R6524-46BPT R6501-26BPT R6502-16BPT R6502-26BPT R6503-26BPT R6504-16BPT R6504-26BPT R6506-26BPT R6508-26BPT R6508-46BPT R650E-26BPT R6510-26BPT R6512-26BPT R6512-46BPT R6516-46BPT R6520-46BPT
224-0550EA 64.6 EUR
224-0550 224-0555 228-0584 224-0562 224-0563 224-0564 224-0566 224-0560 224-0534 224-0535 224-0541 224-0543 224-0546 228-1701 228-1702 228-1689 224-0159 228-1704 228-1705 228-1706 228-1690 228-1707 224-0160 224-0532 224-0537 224-0538 224-0539 224-0540 224-0545 228-0910 228-0635 224-2020 228-1680 228-4319 228-1681 228-1682 228-0634 228-4326 228-4327 224-2028 228-1684 228-4328 228-1685 228-4329 228-4330 228-0400
Pump tubing, PharMed® BPT
Tubazioni Tubo per pompa peristaltica
PharMed® BPT tubing is less permeable to gases and vapours than silicone tubing. It is ideal for cell culture, fermentation, synthesis, separation, purification and process monitoring and control. Independent tests show that PharMed® BPT tubing is safe for use in sensitive cell culture applications.
  • Outlasts silicone tubing in peristaltic pumps by up to 30 times
  • Heat weldable for sterile access in closed systems
  • Documented biocompatibility to the ISO 10993 Standard
PharMed® BPT tubing has very good general chemical resistance and excellent acid,More Product Information