Stampa…
WHEATON® OmniSpence® ELITE, WHEATON® UniSpense® PRO Peristaltic Pump
Fornitore: DWK Life Sciences
W375030-C W375040-C
224-0103EA 4680 EUR
224-0103 224-0104
WHEATON® OmniSpence® ELITE, WHEATON® UniSpense® PRO Peristaltic Pump
Pompe Pompe peristaltiche
The WHEATON® OmniSpence® ELITE and UniSpense® PRO are intelligent peristaltic pumps for all dispensing needs. Whether the user is making sterile liquid transfers to and from containers or filling vials or titer plates these pumps will simplify and accelerate the laboratory throughput. It is ideal for any laboratory or clinical setting where accurate and precise liquid dispensing is important.
  • Highly intuitive icon driven user interface enables fast, easy setup and operation
  • Ultra high precision stepper motor ensures repeatability and accuracy
  • Calibration