Stampa…

Nile blue chloride (Basic blue 12) 85% (dye content), Sigma-Aldrich®

Fornitore: SIGMA ALDRICH MICROSCOPY


M-Clarity™ quality level = MQ200

222550-25G 222550-5G
222550-25GEA 278 EUR
222550-25G 222550-5G
Nile blue chloride (Basic blue 12) 85% (dye content), Sigma-Aldrich®
Nile blue chloride (Basic blue 12)

Nile blue chloride has been used to stain non-aqueous phase in fluorescence microscopy studies.


Nile blue chloride is a green colored powder and is a member of oxazine group of dyes. It changes its color from blue (pH 9,4) to purple-red (11,0). Nile blue chloride has been used to detect DNA and lipids. In addition, it also helps to identify bacterial growth. Nile blue chloride also facilitates diagnosis of disease linked with amyloid accumulation.

Formula: C₂₀H₂₀ClN₃O
Temperatura di stoccaggio: Room Temperature
MDL Number: MFCD00011936
CAS Number: 2381-85-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a636828aaa1ca78fc29a284c5e8143a72da4a496&vid=a636828aaa1ca78fc29a284c5e8143a72da4a496&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance (Colour) Dark Green to Black
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Lambda Max 634 - 642 nm
Carbon Content 56.3 - 79.4%
Nitrogen Content 9.7 - 13.9%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR