Stampa…

D-(+)-Cellobiose, Millipore®

Fornitore: Merck Millipore Calbiochem

Synonyms: β-D-Glc-(1→4)-D-Glc, 4-β-D-glucopyranosyl-D-glueopyranose, Cellose, β-Cellobiose, (2R,3R,4R,5R)-2,3,5,6-Tetrahydroxy-4-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)hexanal, 4-O-β-D-Glucopyranosyl-D-glucopyranose

219458-25
219458-25EA 109 EUR
219458-25
D-(+)-Cellobiose, Millipore®
D-(+)-Cellobiose
Formula: C₁₂H₂₂O₁₁
Peso molecolare: 342.30 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00136034
CAS Number: 528-50-7
EINECS: 208-436-5

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR