Stampa…
Storage Drawers
Fornitore: MULTIROIR
12101MUTBS00A 12102MUTBS02B 12103 12104MUTBS02D
216-4354EA 33.2 EUR
216-4354 216-4362 216-4345 216-4363
Storage Drawers
Scatole Scatole di stoccaggio
Stackable polystyrene drawer unit ideal for organising and storing laboratory components, consumables, and various materials.
  • Can be wall-mounted, fixed to a partition or placed on a countertop
  • Single or double drawer
  • Various sizes available
  • Transparent drawers with label holder
  • Compartmentalised drawers
The modular assembly system makes these pull-out drawers easy to open.