Stampa…
Stainless steel scraper
Fornitore: BURKLE
5386-0030 5386-0035
216-2503EA 91.2 EUR
216-2503 216-2504
Stainless steel scraper
Raschietti
The high-quality scraper is suitable for scraping off, smoothing, cleaning, levelling out and also for removing strongly adhesive, sticky substances or dried up media. The edge of the corrosion-resistant stainless steel scraper is ideal for blending and portioning viscous media.
  • High-grade stainless steel V4A (1.4404), for ideal chemical resistance
  • Extra sturdy design guarantees long service life
  • Sterilisable, autoclavable
The smooth, seamless and sturdy surface ensures cleaning without difficulty and meets hygienic requirements.
The scraper makesMore Product Information