Stampa…
Sample containers, straight, with screw cap, sterile, WHEATON®
Fornitore: DWK Life Sciences
360099 360100 360102 360101 360104 360103 360103PP
216-0992EA 156 EUR
216-0992 216-0994 216-0998 216-0996 216-1010 216-1008 216-1009
Sample containers, straight, with screw cap, sterile, WHEATON®
Contenitori per campioni Recipienti, usi generici
PP translucent or PS clear, with PE or metal screw cap
  • Manufactured from FDA approved virgin polystyrene or polypropylene
  • Supplied sterile and for in vitro use only
  • CE marked in accordance with European Directive 98/79/EC
  • All containers are leak tested to British Standard BS5213:1975(1989)
  • All materials are non cytotoxic
Straight walled vessels ideal for sample collection, storage and transport.
PS offers good thermal and chemical resistance. PP offers excellent thermal and chemical resistance.
Attenzione: *I prodotti in questa pagina non rientranoMore Product Information