Stampa…
Flaconi a collo largo, con tappo a vite, serie 303
Fornitore: Kautex
303-783855 303-783856 303-783859 303-783852
215-9045EA 28.1 EUR
215-9045 215-9046 215-9047 215-9044
Flaconi a collo largo, con tappo a vite, serie 303
Bottiglie Bottiglie in plastica
Polypropylene, round, natural colour.
  • Autoclavable up to 121 °C
  • Hot filled products, pastes, granulates
  • Leakproof
  • Halal certified
Suitable for food.
Note: Unscrew the cap and set on the bottle before autoclaving.
Caps are only suitable for bottles 215-9044 to 215-9047.