Stampa…
Bottles, Narrow Neck, Round
Fornitore: RIXIUS
10200003018OSV 10200010022OSV 10200025022OSV 10200050025OSV 10200005018OSV 10201010022OSV 10201025022OSV 10201050025OSV 10201100028OSV 10501005018OSV 1-0501001518AD 1-0201-0500-45 10201100045OVS 10200100028OSV 10501000518OSV 10501002018OSV 10501003018OSV 10501010018OSV 10201015022OSV 10201020022OSV 10201250045OSV 10500001018XOS 10501001018ADO 1-0200-0125-28
RIXI10200003018OSVEA 168 EUR
RIXI10200003018OSV RIXI10200010022OSV RIXI10200025022OSV RIXI10200050025OSV RIXI10200005018OSV RIXI10201010022OSV RIXI10201025022OSV RIXI10201050025OSV RIXI10201100028OSV RIXI10501005018OSV RIXI1-0501001518AD RIXI1-0201-0500-45 RIXI10201100045OVS 215-4654 215-4655 215-4656 215-4657 215-4658 215-4659 215-4660 215-4661 RIXI10500001018XOS RIXI10501001018ADO RIXI1-0200-0125-28
Bottles, Narrow Neck, Round
Bottiglie Bottiglie in vetro
Clear or amber glass without screw cap.
  • Good chemical and physical properties
  • For applications that do not involve high thermal stresses
  • Round thread in accordance with DIN 168
Excellent for sampling, storing and transporting liquids. Amber coloured to protect light-sensitive materials.