Stampa…
VWR® Bottles, Wide Neck, Square or Rectangular, with Screw Caps
Fornitore: VWR Collection
215-4259EA 81.6 EUR
215-4259
VWR® Bottles, Wide Neck, Square or Rectangular, with Screw Caps
Bottiglie Bottiglie in plastica
Bottles are made with High Density Polyethylene (HDPE) and are available in a natural or amber finish (2,7 L only). Choose from either square or rectangular bottles to meet specific storage requirements.
  • Sterile 10⁻³
  • Wide mouth design for easy filling
  • Graduation marks for reference during sample collection
  • Packed in smaller packs for better handling and to reduce contamination
  • CE IVD Directive 98/79/EC
Ideal for the storage of liquids, solids, and powders. These bottles are generally used for large volume urine sample collection. Bottles supplied withMore Product Information