Stampa…
Sample vials, E-C, WHEATON®
Fornitore: DWK Life Sciences
224722 224724 224706 224741 224711 224712 224714 224728 224721 224740 224750 224744 224708 224701 224705 224710 224742 224720 224725 224726 224730 224731 224732 224745 224748 224752 224734 W224810 W226060
215-3572EA 243 EUR
215-3572 215-3573 218-2261 218-2265 218-2266 218-2267 218-2268 215-3576 215-3571 WHEA224740 WHEA224750 215-3578 WHEA224708 WHEA224701 WHEA224705 WHEA224710 215-3577 WHEA224720 215-3574 215-3575 WHEA224730 WHEA224731 WHEA224732 215-3579 215-3581 WHEA224752 WHEA224734 WHEAW224810 WHEAW226060
Sample vials, E-C, WHEATON®
Vial
Clear vials made from low extractable borosilicate glass that provides superior chemical resistance. Conform to USP Type I and ASTM E 438 Type I, Class A requirements.
Amber vials made from borosilicate glass that conforms to USP Type I requirements for light transmission to protect light-sensitive products.
Informazioni per l'ordine: Caps and replacement caps can be purchased separately.
Confezione: Vials packed in two shrink-wrapped packs of 100 vials which reduces contamination from corrugate. The 60 ml vials are packed in one shrink-wrapped pack of 143 vials.