Stampa…
Ring weights for vessels, SP Bel-Art
Fornitore: Bel-Art Products, a Part of SP
F183070003 F183070005 F183070010 F183070015 F183070020
214-1941EA 64.3 EUR
214-1941 214-0170 214-0171 214-0172 214-0173
Ring weights for vessels, SP Bel-Art
Pesi Pesi per matracci
Lead core, with Vikem® vinyl coating.
  • Bright red, non-slip Vikem® coating prevents damage to surfaces
  • Good chemical resistance
  • Stabilise glassware immersed in water baths; reduce floating, breakages and chemical spills