Stampa…
Disposable culture tubes
Fornitore: Simport Scientific
T400-10A T400-3A T400-3 T400-3B T400-3G T400-3Y T400-3O T400-3AB T400-3AG T400-3AY T400-3AO T400-4 T400-10 T400-7 T400-3DS T400-3ADS T400-3ALST T400-4A T400-3S T400-3AS T400-3AAM
212-9597EA 186 EUR
212-9597 212-9599 212-9598 212-9603 720-0544 212-9604 720-0545 SIMPT400-3AB 212-9600 212-9602 212-9601 212-9605 212-9596 SIMPT400-7 SIMPT400-3DS SIMPT400-3ADS SIMPT400-3ALST SIMPT400-4A SIMPT400-3S SIMPT400-3AS SIMPT400-3AAM
Disposable culture tubes
Tubi Provette per colture
PS o PP
  • Manufactured without the use of release agents that could cause errors and interference in RIA tests
  • Precision moulding ensures uniform size and shape
  • PP tubes are translucent, can withstand centrifugation speeds over 3000×g, are suitable for most common acids, solvents and alkalis at room temperature, are almost unbreakable, and can be sterilised at 120 °C
  • PS tubes are transparent, will withstand centrifugation speeds up to 1400×g, can tolerate aqueous solutions, mild bases and weak acids (but not organic solvents, aromatic or chlorinated