Stampa…
Test tubes, without rim, medium wall, `SAMCO 50`
Fornitore: MURRAY S
G023/31 G023/32 G023/33 G023/35 G023/36 G023/38 G023/40 G023/34
MURRG023/31EA 40.7 EUR
MURRG023/31 MURRG023/32 MURRG023/33 212-0457 212-0458 212-0459 212-0460 MURRG023/34
Test tubes, without rim, medium wall, `SAMCO 50`
Tubi Provette
Test tubes, made of SAMCO neutral glas.
Neutral glass test tubes will withstand moderate temperature change, they should not be taken from cold and placed directly into the hottest part of the flame without pre-heating.