Stampa…
VWR® PCR Plates, 96-Well
Fornitore: VWR Collection
211-0262EA 323 EUR
211-0262 211-0263 211-0264 211-0265 211-0266 211-0267 211-0268 211-0269 211-0298 211-0313 211-0283 732-3487 732-3488 732-3494
VWR® PCR Plates, 96-Well
Micropiastre Piastre per PCR
VWR® 96-Well PCR Plates are compatible with most leading PCR and real-time PCR instruments.
  • Ultra-thin walls to optimize thermal transfer resulting in increased reaction efficiencies
  • Ultra-clear wells for easy visualization of small sample volumes
  • Made of virgin polypropylene which is naturally low-binding without the need to incorporate additives
  • Black alphanumerics are easy-to-read and simplifies sample identification.
  • All plates are certified free of RNAse, DNAse, and human DNA
Compatible with VWR® 8-strip flat caps (211-0350) and bubble caps (732-0More Product Information