Stampa…

Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi

Fornitore: VWR Chemicals
Pericolo

Synonyms: Ethyl alcohol

20823.293EA 32.6 EUR
20823.293 20823.327 20823.362 20823.555 20823.293DP 20823.327DP 20823.362DP 20823.460 20823.460DP 20823.290 VWRC20823.290DP
Etanolo 96% (v/v), AnalaR NORMAPUR® per analisi
Etanolo

Attenzione: ATTENZIONE: questo alcol e regolamentato. Vi preghiamo di contattare i nostri uffici.

Formula: C₂H₅OH
Peso molecolare: 46.07 g/mol
Punto di ebollizione: 78.3 °C (1013 hPa)
Punto di fusione: –117 °C
Densità: 0.7895 g/cm³ (20 °C)
Flash Pt: 12 °C (coppa chiusa)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003568
CAS Number: 64-17-5
EINECS: 200-578-6
UN: 1170
ADR: 3 II
REACH: 01-2119457610-43
Merck Index: 12,03806

Visualizza i codici

Risultati del test di specificazione

Assay (on anhydrous substance) Min. 99.8 %
Assay (V/V) 96 - 96,3 %
IR Spectrum Passes test
UV Spectrum Passes test
Acidity Max. 0.0002 meq/g
Alkalinity Max. 0.0001 meq/g
Boiling point 78.0 - 78.5 °C
Colouration Max. 10 APHA
Substances coloured by H2SO4 Max. 20 APHA
Aldehydes (as CH3CHO) Max. 5 ppm
Aldehydes + ketones (as CH3CHO) Max. 10 ppm
Benzene Max. 5 ppm
Evaporation residue Max. 5 ppm
Formaldehyde Max. 1 ppm
Furaldehyde Max. 1 ppm
Ketones (as CH3COCH3) Max. 10 ppm
Methanol Max. 0.02 %
Propan-2-ol Max. 100 ppm
Substances reducing KMnO4 (as O) Max. 4 ppm
Total heavy alcohols Max. 100 ppm
Water 5.2 - 8.2 %
Al (Aluminium) Max. 0.1 ppm
Ba (Barium) Max. 0.02 ppm
Ca (Calcium) Max. 0.2 ppm
Cd (Cadmium) Max. 0.02 ppm
Co (Cobalt) Max. 0.01 ppm
Cr (Chromium) Max. 0.01 ppm
Cu (Copper) Max. 0.1 ppm
Fe (Iron) Max. 0.05 ppm
K (Potassium) Max. 0.1 ppm
Mg (Magnesium) Max. 0.05 ppm
Mn (Manganese) Max. 0.01 ppm
Na (Sodium) Max. 1 ppm
Ni (Nickel) Max. 0.02 ppm
Pb (Lead) Max. 0.05 ppm
Sn (Tin) Max. 0.05 ppm
Sr (Strontium) Max. 0.02 ppm
Zn (Zinc) Max. 0.1 ppm

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR