Stampa…

Acido nitrico 60%, AnalaR NORMAPUR® zA

Fornitore: VWR Chemicals
Pericolo

Synonyms: Nitric acid solution

20423.291EA 58.8 EUR
20423.291 20423.360
Acido nitrico 60%, AnalaR NORMAPUR® zA
Acido nitrico
Formula: HNO₃
Peso molecolare: 63.01 g/mol
Punto di ebollizione: 87
Punto di fusione: -42
Densità: 1.51
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011349
CAS Number: 7697-37-2
EINECS: 231-714-2
UN: 2031
ADR: 8 II
Merck Index: 13,06610

Visualizza i codici

Risultati del test di specificazione

Assay Min. 60.0 %
Density (20/4) 1.365 - 1.375
Heavy metals (as Pb) Max. 0.2 ppm
Ignition residue (SO4) Max. 10 ppm
Cl (Chloride) Max. 0.5 ppm
PO4 (Phosphate) Max. 2 ppm
SO4 (Sulphate) Max. 2 ppm
As (Arsenic) Max. 0.01 ppm
Fe (Iron) Max. 1 ppm
Mn (Manganese) Max. 0.4 ppm
Pb (Lead) Max. 0.05 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR