Stampa…
Sorbent tubes for gas and vapor sampling, ORBO™
Fornitore: Merck
20267-U 20257-U 20265 20349 20351 20355 20358 20360 20362 20832-U 20030-U 20044 20051 20053 20063 20081-U 20211 20254-U 20255-U 20256-U 54020-U 23446-U 54081-U
20267-UEA 135 EUR
20267-U 20257-U SUPL20265 20349. SUPL20351 20355. 20358. SUPL20360 20362. 20832-U 20030-U SUPL20044 SUPL20051 SUPL20053 SUPL20063 20081-U SUPL20211 20254-U 20255-U 20256-U 54020-U 23446-U 54081-U
Sorbent tubes for gas and vapor sampling, ORBO™
Fiale per gas
ORBO™ sorbent tubes contain two beds of the same selective adsorbent seperated by glass wool or foam, for gas and vapor sampling.
  • Wide range of high quality adsorbents
  • Consistent adsorbent mesh sizes
  • Coated and specially-treated materials
  • Reproducible recoveries and low standard deviation
  • Consistent bed weights from tube to tube to ensure capacity
The dual-layer or two-bed construction of the tube allows for any sample breakthough to be captured in the smaller back-up bed.