Stampa…
Anti-IgG Chicken Polyclonal Antibody (CF™ 543)
Fornitore: Biotium
20335-50UL
20335-50ULEA 121 EUR
20335-50UL
Anti-IgG Chicken Polyclonal Antibody (CF™ 543)
Anticorpi
CF™543 antibodies are affinity-purified antibodies labeled with orange fluorescent CF™543 (Ex/Em 541/560nm), a member of the novel CF™ dye series developed by Biotium for labeling proteins and other biomolecules. CF™ dyes are designed to offer advantages in brightness, photostability, and/or specificity compared to other commercial dyes, and are available in colors spanning the visible, far-red, and near-infrared spectra.
CF™543 is an orange fluorescent dye with an excitation spectrum optimally matching the 543 laser line. Antibody conjugates of the dye are much brighter than those ofMore Product Information