Stampa…

Anti-IgG Goat Polyclonal Antibody (CF™ 568)

Fornitore: Biotium
20104 20104-1
20104.EA 359 EUR
20104. 20104-1
Anti-IgG Goat Polyclonal Antibody (CF™ 568)
Anticorpi
Type: Secondary
Antigen: IgG (Chicken) (chicken IgG)
Clonality: polyclonal
Clone:
Conjugation: CF® 568 Dye
Epitope:
Host: Goat
Isotype: none
Reactivity: Chicken
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR