Stampa…
Flasks, iodine with collar and iodine stopper
Fornitore: Lenz Laborglas GmbH & CO.KG
03026249
201-1162EA 78.9 EUR
201-1162
Flasks, iodine with collar and iodine stopper
Fiasche Beute per numero di iodio
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Volume graduations and labelling area.