Stampa…
Fractionating columns, Vigreux
Fornitore: witeg Labortechnik
1.521.200 1.522.300 1.523.600 1.521.300 1.523.300 1.521.201 1.520.201 1.523.201
201-0175EA 91.4 EUR
201-0175 201-0177 201-0179 201-0176 201-0178 WITG1.521.201 WITG1.520.201 WITG1.523.201
Fractionating columns, Vigreux
Componenti per distillazione Colonne di distillazione
Borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
With or without glass jacket.