Stampa…

1-Butylboronic acid, Supelco®

Fornitore: Merck

Synonyms: Butylboronic acid, n-Butylboronic acid

M-Clarity™ quality level = MQ100

19667-5G
19667-5GEA 301 EUR
19667-5G
1-Butylboronic acid, Supelco®
1-Butylboronic acid
Formula: C₄H₁₁BO₂
MDL Number: MFCD00002106
CAS Number: 4426-47-5
EINECS: 224-607-7

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR