Stampa…

D(+)-Glucosio monoidrato, in polvere, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®

Fornitore: Avantor

Synonyms: (2S,3R,4S,5S,6R)-6-(Hydroxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetraol hydrate, Dextrose monohydrate

1910-05EA 381.92 EUR
1910-05
D(+)-Glucosio monoidrato, in polvere, BAKER ANALYZED®, Laboratory Reagent, J.T.Baker®
D(+)-Glucosio monoidrato
Formula: C₆H₁₂O₆·H₂O
Peso molecolare: 198.17 g/mol
Punto di fusione: 83
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149450
CAS Number: 14431-43-7
EINECS: 200-075-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c9ea2af2ca62ca94d86809070593a60aafd8cd1b&vid=e549b499dca7fb95811640efad1429268c8f6d8b&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

For Laboratory,Research,or Manufacturing Use = 0.005 %
Insoluble Matter ≤ 0.005 %
Residue after Ignition ≤ 0.015 %
Chloride (Cl) ≤ 0.005 %
Sulfate and Sulfite (as SO₄) ≤ 0.005 %
Starch Passes Test
Arsenic (As) ≤ 0.200 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤ 5 ppm
Trace Impurities - Iron (Fe) ≤ 5 ppm


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR