Stampa…

Argento carbonato ∼ 0,7 mmole Ag₂CO₃/g on Celite

Fornitore: Thermo Fisher Scientific
Attenzione

18202.W5 18202.W9
18202.W5EA 208 EUR
18202.W5 18202.W9
Argento carbonato ∼ 0,7 mmole Ag₂CO₃/g on Celite
Argento carbonato

∼0.7 mmole Ag₂CO₃/g reagent


CAS No.: 534-16-7 Molecular Formula: Ag2CO3/Celite Physical Form: Powder Sensitivity: Light Sensitive Odour: Odourless MDL No.: MFCD00003403


Oxidizing agent for alcohols, diols/triols, hydroquinones, and amines.

Formula: CAg₂O₃
Punto di fusione: 210…218 °C
Densità: 5,65 g/cm³ (20 °C)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00003403
CAS Number: 534-16-7
EINECS: 208-590-3
UN: 3077
ADR: 9,III
Merck Index: 12,08650

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR