Stampa…

Apigenin, Millipore®

Fornitore: Merck Millipore Calbiochem

178278-5
178278-5EA 97.3 EUR
178278-5
Apigenin, Millipore®
Apigenin
Formula: C₁₅H₁₀O₅
MDL Number: MFCD00006831
CAS Number: 520-36-5
EINECS: 208-292-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR