Stampa…

Bismuth(III) bromide ≥99% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo

17112.14 17112.22
17112.14EA 125 EUR
17112.14 17112.22
Bismuth(III) bromide ≥99% (metals basis)
Bismuth(III) bromide

Purity: 99% CAS No.: 7787-58-8 Molecular Formula: BiBr3 Formula Weight: 448.71 Physical Form: Powder Density: 5.72 Appearance: Yellow Odour: Acidic Melting Point: 218° Boiling Point: 453° UN No.: 3260 MDL No.: MFCD00014195

Formula: BiBr₃
Punto di ebollizione: 453 °C
Punto di fusione: 218 °C
Densità: 5,72
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00014195
CAS Number: 7787-58-8
EINECS: 232-121-1
UN: 3260
ADR: 8,III
Merck Index: 12,01299

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR