Stampa…
Sistemi per acqua ultrapura, arium® pro
Fornitore: Sartorius
H2OPRO-DI-T H2OPRO-UF-B H2OPRO-UF-T H2OPRO-UV-B H2OPRO-UV-T H2OPRO-VF-B H2OPRO-VF-T H2OPRO-DI-B H2OPRO-UV-BTOC H2OPRO-UV-TTOC H2OPRO-VF-BTOC H2OPRO-VF-TTOC
171-0394EA 7720 EUR
171-0394 171-0382 171-0384 171-0385 171-0387 171-0388 171-0390 171-0648 171-0558 171-0557 171-0561 171-0560
Sistemi per acqua ultrapura, arium® pro
Purificatori d'acqua Sistemi per acqua pura e ultrapura
Arium® pro Type I water purification systems are designed for sophisticated users to meet all their requirements for reagent grade water, needed both for critical applications and routine analyses. Different versions are available for reliable and optimal analytical results.
  • Compact design, flexibility of use and simplicity of maintenance
  • Innovative glass operator interface with touch function
  • Service and alarm functions supported by graphic displays
  • Conductivity measurement for feed and product water
  • Integrated monitoring of water consumption
  • RNases: <0