Stampa…
Affinity coupling media, EAH Sepharose™ 4B
Fornitore: Cytiva
17-0569-01
17-0569-01EA 615 EUR
17-0569-01
Affinity coupling media, EAH Sepharose™ 4B
Purificazione proteine Resine per la purificazione delle proteine
EAH Sepharose™ 4B allows for the stable carbodiimide-based coupling of carboxyl groups to Sepharose® 4B via an 11-atom hydrophilic spacer arm that permits extremely stable coupling.
Stable carbodiimide-based coupling of carboxyl groups to 4B via an 11-atom hydrophilic spacer arm permits very stable coupling. Stable to all commonly used aqueous Sepharose buffers.