Stampa…
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Fornitore: Cytiva
17-0230-01
17-0230-01EA 475 EUR
17-0230-01
SP Sephadex™ C-25 Ion Exchange Chromatography Media, Cytiva
Purificazione proteine Resine per la purificazione delle proteine
SP Sephadex™ C-25 is a strong cation exchanger. The ion exchange group is a sulphopropyl group which remains charged and maintains consistently high capacity over the entire working range of pH 4 to 13.
The ion exchange group is a sulphopropyl group which remains charged and maintains consistently high capacity over the entire working range of pH 4 to 13.