Stampa…

Boron trifluoride - methanol complex 10% in methanol, Supelco®

Fornitore: Merck
Pericolo


M-Clarity™ quality level = MQ100

15716-100ML 15716-10ML 15716-1L 15716-250ML 15716-1ML
15716-100MLEA 63.8 EUR
15716-100ML 15716-10ML 15716-1L 15716-250ML 15716-1ML
Boron trifluoride - methanol complex 10% in methanol, Supelco®
Boron trifluoride - methanol complex
Formula: CH₄BF₃O
MDL Number: MFCD00071635
CAS Number: 373-57-9
EINECS: 206-766-4
UN: 3286
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=612f76cb6db91dcf7502c059b61da9608e7730d7&vid=9a47800cc30d1a2cd0d48bc41fdacc864dfce83f&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR