Stampa…

Lead ionophore IV, Selectophore™ ionophore, Supelco®

Fornitore: Merck

Synonyms: tert-Butylcalix[4]arene-tetrakis(N,N-dimethylthioacetamide)

MQ100

15343-50MG
15343-50MGEA 245 EUR
15343-50MG
Lead ionophore IV, Selectophore™ ionophore, Supelco®
Lead ionophore IV

Lead ionophore IV (tert-Butylcalix[4]arene-tetrakis(N,N-dimethylthioacetamide)), was used in the fabrication of robust potentiometric ion sensors


It was also used as the base for the synthesis of lead-selective micro-electrode.


Confezione: Bottomless glass bottle. Contents are inside inserted fused cone.

Formula: C₆₀H₈₄N₄O₄S₄
Peso molecolare: 1053.59 g/mol
MDL Number: MFCD00679123
CAS Number: 145237-46-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49feb73527b2af4562e0d6a12f3a1dfeebd8c388&vid=20a8e30822df94bb1df8288dad019ebed2d3c3cf&prid=web&clid=SAPDAM

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR