Stampa…

Nichel (II) cloruro, anidro ≥99% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo

Synonyms: Dicloruro di nichel

14687.22 14687.36
14687.22EA 121 EUR
14687.22 14687.36
Nichel (II) cloruro, anidro ≥99% (metals basis)
Nichel (II) cloruro

Purity: 99% CAS No.: 7718-54-9 Molecular Formula: Cl2Ni Formula Weight: 129.60 Physical Form: Powder Density: 3.55 Appearance: Orange Odour: Odourless Melting Point: 1001° Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00011142

Formula: Cl₂Ni
Punto di fusione: 1001 °C
Densità: 3,55
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011142
CAS Number: 7718-54-9
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,6505

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR