Stampa…

Moxifloxacin Hydrochloride, Reference Standard

Fornitore: USP

1448606
1448606.EA 944 EUR
1448606.
Moxifloxacin Hydrochloride, Reference Standard
Standard Standard organici

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certificazioni: US Pharmacopea Reference Standard

Visualizza i codici

Specifiche

Descrizione Moxifloxacin Hydrochloride
Environmentally Preferable
CAS No. 192927-63-2
Formula C₂₁H₂₅ClFN₃O₄

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR