Stampa…

Montelukast, Reference Standards

Fornitore: USP

1446815 1446837 1446848 1446859
1446815.EA 1230 EUR
1446815. 1446837. 1446848. 1446859.
Montelukast, Reference Standards
Standard Standard organici

USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.


Certificazioni: US Pharmacopea Reference Standard

Visualizza i codici

Specifiche

Descrizione Montelukast Dicyclohexylamine Montelukast racemate Montelukast Sodium
Environmentally Preferable
CAS No. 577953-88-9 577953-88-9 151767-02-1
Formula C₄₇H₅₉ClN₂O₃S C₃₅H₃₅ClNNaO₃S

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR