Stampa…

Piombo (II) nitrato ≥99.0% ACS

Fornitore: Thermo Fisher Scientific
Pericolo

14243.36 14243.A3
14243.36EA 54 EUR
14243.36 14243.A3
Piombo (II) nitrato ≥99.0% ACS
Piombo (II) nitrato

Purity: 99.0% min CAS No.: 10099-74-8 Molecular Formula: Pb(NO3)2 Formula Weight: 331.20 Physical Form: Crystalline Density: 4.530 Appearance: White Odour: Odourless Melting Point: 470° dec. Solubility: Freely soluble in water. Soluble in absolute alcohol, methanol. Insoluble in concentrated HNO3 Merck Reference: 14,5414 UN No.: 1469 MDL No.: MFCD00011153


As an oxidizer in dye industry, in manufacturing of mother-of-pearl, as mordant in printing textile, as sensitizer in photography

Formula: Pb(NO₃)₂
Punto di fusione: 470 °C (dec.)
Densità: 4,530
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011153
CAS Number: 10099-74-8
EINECS: 233-245-9
UN: 1469
ADR: 5.1,II
Merck Index: 14,5414

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR