Stampa…
SlimLine PLW 7111 Laboratory Glasswashers
Fornitore: MIELE
11882110 11882140 11882150 11882170 11882190 11882230 11882240
141-1606EA 37155 EUR
141-1606 141-1607 141-1608 141-1609 141-1610 141-1611 141-1612
SlimLine PLW 7111 Laboratory Glasswashers
Lavavetreria
The PLW 7111 SlimLine laboratory washers offer a comprehensive and systematic solution for analytically pure reprocessing of laboratory glassware.
  • Smart Load Plus - for flexible use of wash cabinet space
  • EasyLoad - systematic efficiency and convenience
  • Space-saving SlimLine design
  • Freely programmable controls
  • Flexible reprocessing of laboratory glassware
  • Status display via chamber lighting
  • Connectivity-ready
The PLW 7111 series of SlimLine laboratory glasswashers provide performance and user-friendliness. This entirely redesigned washing system boastsMore Product Information