Stampa…
Industrial vacuum cleaner, AS400 HEPA
Fornitore: TIGER-VAC
110766B
141-1548EA 1390 EUR
141-1548
Industrial vacuum cleaner, AS400 HEPA
Aspirapolvere
This vacuum cleaner made of heavy duty plastic, is designed for the recovery of lead, asbestos, silica and other toxic/nuisance dusts.
  • 3-stage filtration system which consists of a HEPA Filter, main filter, disposable filter bag
  • Lightweight and portable
  • By-pass heavy duty motor
  • Heavy duty plastic recovery tank (stainless steel recovery tank available on request)
  • Ready to use: filter and tool kit included
  • Mounted on a a 4-wheel dolly for easy maneuverability
  • Built-in tool storage tray with two large rear wheels (170 mm) and two front wheels (60 mm)