Stampa…

Lithium cobaltite (lithium cobalt(III) oxide) ≥97%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: Lithium cobalt(III) oxide

14049.18 14049.30 14049.A1
14049.18EA 78.7 EUR
14049.18 14049.30 14049.A1
Lithium cobaltite (lithium cobalt(III) oxide) ≥97%
Lithium cobaltite (lithium cobalt(III) oxide)

Purity: 97% CAS No.: 12190-79-3 Molecular Formula: LiCoO2 Formula Weight: 97.88 Physical Form: Powder Sensitivity: Hygroscopic MDL No.: MFCD00049786

Formula: CoLiO₂
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00049786
CAS Number: 12190-79-3
EINECS: 235-362-0

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR