Stampa…
Chrome security truck, Super Erecta®
Fornitore: Metro International
SEC56DC SEC53DC SEC55EC SEC53EC SEC56EC SEC66EC
139-0023EA 2660 EUR
139-0023 139-0804 139-0367 139-0366 139-0368 METOSEC66EC
Chrome security truck, Super Erecta®
Arredi Ripiani
The chrome finish mobile security standard-duty stem caster units protect valuable materials and sensitive items from loss and they are ideal for use in dry environments.
  • The heavy-gauge open wire construction keeps contents visible at all time making it easy to check inventory
  • The units are provided with casters and the shelves can be positioned in 25 mm increments along the entire height of post
  • Double doors open 270 degrees and can be secured along the sides of the unit
Informazioni per l'ordine: The intermediate shelves are not included with baseMore Product Information