Stampa…

Idruro di litio-alluminio ≥95%

Fornitore: Thermo Fisher Scientific
Pericolo

13442.14 13442.22 13442.36 13442.51
13442.14EA 52.2 EUR
13442.14 13442.22 13442.36 13442.51
Idruro di litio-alluminio ≥95%
Idruro di litio-alluminio

Purity: 95% min CAS No.: 16853-85-3 Molecular Formula: LiAlH4 Formula Weight: 37.95 Physical Form: Powder Density: 0.917 Appearance: White to grey Odour: Odourless Melting Point: 125° dec. Solubility: Decomposes in water. Soluble in ether Merck Reference: 14,349 UN No.: 1410 MDL No.: MFCD00011075


Reducing agent for pharmaceuticals, perfumes, and fine organic chemicals; source of hydrogen, propellant, and catalyst in polymerizations

Formula: H₄AlLi
Punto di fusione: >125 °C (dec.)
Densità: 0,917
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011075
CAS Number: 16853-85-3
EINECS: 240-877-9
UN: 1410
ADR: 4.3,I
Merck Index: 14,349

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR