Stampa…

Tungsteno triossido ≥99.998% (metals basis excluding Mo), Puratronic® (max. 0.01% Mo)

Fornitore: Thermo Fisher Scientific
Attenzione

13398.14 13398.22
13398.14EA 176 EUR
13398.14 13398.22
Tungsteno triossido ≥99.998% (metals basis excluding Mo), Puratronic® (max. 0.01% Mo)
Tungsteno triossido

Purity: 99.998% CAS No.: 1314-35-8 Molecular Formula: O3W Formula Weight: 231.85 Physical Form: Powder Density: 7.16 Melting Point: 1473° Merck Reference: 14,9817 MDL No.: MFCD00011466

Formula: O₃W
Punto di fusione: 1473 °C
Densità: 7,16
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011466
CAS Number: 1314-35-8
EINECS: 215-231-4
Merck Index: 14,9817

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR