Stampa…
Diaphragm pumps, VARIO® select
Fornitore: VACUUBRAND
20736350 20738150
131-0033EA 6980 EUR
131-0033 131-0050
Diaphragm pumps, VARIO® select
Pompe Pompe da vuoto a membrana
The VACUU·SELECT controller simplifies your work and allows VARIO select diaphragm pumps to deliver unparalleled precision. The pumps run whisper quiet and respond to demand in real time by matching pumping speed to process conditions. The intuitive VACUU·SELECT controller includes predefined applications for all common vacuum processes. VACUUBRAND VARIO® technology - frees you up for other tasks.
  • VACUU·SELECT vacuum controller with graphical user interface and predefined applications simplifies work in the laboratory
  • Contamination-free pumping and