Stampa…
Doypack® Paper/PET/CPP Paper Bags
Fornitore: Flexico
ITM080351 ITM080352 ITM080353 ITM080354 ITM080355
129-3901EA 230 EUR
129-3901 129-3902 129-3903 129-3904 129-3905
Doypack® Paper/PET/CPP Paper Bags
Sacchetti Sacchetti di stoccaggio
The specific shape of the Doypack® bags allows a good vertical position of the packaging on the shelves. The Paper/PET/CPP laminate provides a good barrier to gases and humidity. The paper layer gives the bag a natural look.
  • Food contact
  • Available in different sizes
  • Ideal for wet and dry content
  • Doypack® packaging
  • Ideal for shelf display
  • Available in stock
The Doypack® packaging has a window to give a better visibility of the content and highlights the features of the product. Ideal for food products (biscuits, confectionery, snacks) this bag isMore Product Information