Stampa…
Sacchetti per autoclave
Fornitore: Greiner Bio-One
643201_500 646201_500 649201_350 645201_500 BAG1_200 BAG2_200 BAG3_200
129-1750EA 216 EUR
129-1750 129-1752 129-1753 GREI645201_500 129-1754 129-1755 129-1756
Sacchetti per autoclave
Sacchetti Sacchetti smaltimento rifiuti
These disposal bags are used for storing contaminated petri dishes or test tubes.
  • Suitable for autoclaving by wet steam
  • Specially designed to eliminate contaminated waste in the laboratory
PP, transparent, thickness: 50 µm