Stampa…
VWR®, Incineration Bags
Fornitore: VWR Collection
129-1567EA 220 EUR
129-1567 129-1569
VWR®, Incineration Bags
Sacchetti Sacchetti smaltimento rifiuti
Constructed of durable PP with feature the universal biohazard symbol in black.
Certificazioni: Meets the 165 g dart drop standard performed per ASTM D1709-98 and the 480 g tear standard performed per ASTM D1299-94A.
Attenzione: Autoclaving not recommended. Bags will melt at 98,9 °C.