Stampa…
Sterilised standard bags, Whirl-Pak®
Fornitore: NASCO FORT ATKINSON
B01009WA B00679WA B00736WA B01020WA B00990WA B00994WA B01027WA B01018WA B00992WA B01323WA B01063WA B01532WA
432-0166EA 146 EUR
432-0166 432-0162 129-0667 129-0668 432-0163 432-0165 432-0168 432-0167 432-0164 129-0670 129-0672 NASCB01532WA
Sterilised standard bags, Whirl-Pak®
Sacchetti Sacchetti per campioni
Blended, virgin polyethylene film provides exceptional clarity, superior strength, and sterilised after manufacturing for results you can trust. HACCP, EPA, USDA, FDA approved for food contact.
  • Plus 1 sterilisation, post-manufacturing sterilization: Increased sample integrity, perforated closure to maintain sterility
  • Puncture-proof closure tabs: Inner and outer tapes for longevity and stability, wire for easy opening, pouring, and closing
  • Opening pull tabs: For easy opening, non-slip textured Tyvek
  • Continuous perimeter seal: Maximum protection against