Stampa…

Zinc(II) acetylacetonate monohydrate

Fornitore: Thermo Fisher Scientific
Attenzione

12527.18 12527.30
12527.18EA 36.4 EUR
12527.18 12527.30
Zinc(II) acetylacetonate monohydrate
Zinc(II) acetylacetonate monohydrate

CAS No.: 14363-15-6 Molecular Formula: C10H14O4Zn.H2O Formula Weight: 281.60 (263.59anhy) Physical Form: Crystalline Density: 1.6 Melting Point: 136-138° dec. MDL No.: MFCD00000035

Formula: C₁₀H₁₄O₄Zn·1H₂O
Punto di fusione: 136…138 °C (dec.)
Densità: 1,6
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000035
CAS Number: 14363-15-6
EINECS: 237-860-3

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR