Stampa…

Nichel (II) cloruro esaidrato ≥99.3% (metals basis)

Fornitore: Thermo Fisher Scientific
Pericolo

12372.A3 12372.36
12372.A3EA 180 EUR
12372.A3 12372.36
Nichel (II) cloruro esaidrato ≥99.3% (metals basis)
Nichel (II) cloruro esaidrato

Purity: 99.3% CAS No.: 7791-20-0 Molecular Formula: Cl2Ni.6H2O Formula Weight: 237.71 (129.60anhy) Physical Form: Crystalline Density: 1.92 Appearance: Green Odour: Odourless pH: 4.9 Melting Point: 140° -H2O Merck Reference: 14,6505 UN No.: 3288 MDL No.: MFCD00149809

Formula: NiCl₂·6H₂O
Punto di fusione: 140 °C (-H₂O)
Densità: 1,92
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00149809
CAS Number: 7791-20-0
EINECS: 231-743-0
UN: 3288
ADR: 6.1,III
Merck Index: 14,6505

Visualizza i codici

Specifiche

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR